missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPA2 Monoclonal Antibody (OTI1C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504622
This item is not returnable.
View return policy
Description
Three different forms of human pancreatic procarboxypeptidase A have been isolated. The encoded protein represents the A2 form, which is a monomeric protein with different biochemical properties from the A1 and A3 forms. The A2 form of pancreatic procarboxypeptidase acts on aromatic C-terminal residues and is a secreted protein.Specifications
| CPA2 | |
| Monoclonal | |
| 0.89 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P48052 | |
| CPA2 | |
| Full length human recombit protein of human CPA2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C7 | |
| Unconjugated | |
| CPA2 | |
| carboxypeptidase A2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1358 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction