Learn More
CPA2 Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504645
Encompass_Preferred
 View more versions of this product
Product Code. 17502122
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Three different forms of human pancreatic procarboxypeptidase A have been isolated. The encoded protein represents the A2 form, which is a monomeric protein with different biochemical properties from the A1 and A3 forms. The A2 form of pancreatic procarboxypeptidase acts on aromatic C-terminal residues and is a secreted protein.
Specifications
Specifications
CPA2
Monoclonal
1.11 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P48052
CPA2
Full length human recombit protein of human CPA2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1D8
Unconjugated
CPA2
carboxypeptidase A2
Mouse
Affinity Chromatography
RUO
1358
-20° C, Avoid Freeze/Thaw Cycles