Learn More
Cole-Parmer™ CPC™ Metal Quick Disconnect Straight-through Compression Fitting Insert
Ideal for use with gases or liquids
Merk:  Cole-Parmer™ 31404-48
Overige gegevens : Gewicht : 0.01000kg
 Bekijk meerdere types van dit product via de
Produktcode. 15447253
  • € 8.35 / 1 stuk
onze contactgegevens 020 4877000
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately).
  • Withstands vacuum to 250psi.
  • Temperature range: -40 to +180°F.
Specificatie
Specificatie
Quick-Disconnect x Compression Connection
Brass
Quick-disconnect Compression Fitting Insert
82°C
-40°C
1/8 in.
Yes
4.37cm
250psi
-40°F
5/32 in.
Autoclave or Ethylene Oxide (ETO)
Video's