Learn More
Cole-Parmer™ CPC™ NSH Non-spill Coupling Insert with Hose Barb
Click to view available options
Tubing Size:
1/2 in.
Unit Size:
Each
Description
- NS6 Series couplings feature non-spill valves at a great price.
- Use the NS6 when even a few drops of spillage pose problems regarding safety, media cost or environmental regulations.
- Soft touch overmold makes these couplings comfortable in the hand and very attractive.
- Lightweight, chemically resistant and easy to use.
- Non-spill design.
- Disconnect under pressure with no spills.
- Color coding Instant visual differentiation of media lines.
- Glass-filled polypropylene.
- Durable and compatible with many chemicals.
- Colder thumb latch.
- One-hand connection and disconnection.
Specifications
Specifications
| Description | Quick-Disconnect x Barb |
| Material | Polypropylene |
| Tubing Size | 1/2 in. |
| Hose Barb | 1/2 in. |
| Outlet Port | 1/2 in. Quick Disconnect |
| Product Type | Non-Spill, Quick-Disconnect x Compression Coupling Insert |