Learn More
Cole-Parmer™ CPC™ Quick Disconnect Polypropylene Pipe Adapter Bodies
Click to view available options
Thread Size:
1/2 in.
3/4 in.
Unit Size:
Each
Description
- High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately)
- Plug insert into coupling body to connect
- Fitting halves lock into place immediately
- To disconnect fitting halves, simply press the button on the coupling body
- Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged
- All fitting bodies have 316 SS latch springs and EPDM seals
Specifications
Specifications
| Type | Adapter |
| Temperature (English) Max. | 160°F |
| Temperature (Metric) Max | 71°C |
| Thread Size | 1/2 in. |
| Port Type | 1/2 in. Quick-Disconnect |
| Description | Quick-Disconnect x NPT(M) Connection |
| Sterilization Method | Ethylene Oxide (ETO) or Gamma |
| Material | Polypropylene |
| Pressure | 60 psi |