Learn More
Cole-Parmer™ CPC™ Sanitary Connector, PC Sleeve, Single-Cycle
Product Code. 15497373
Click to view available options
Tubing Size:
3/4 in.
3/4 in. x 1-1/2 in.
Unit Size:
Each
Product Code. 15497373
Brand: Cole-Parmer™ 3160422
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for media manufacturing, cell harvesting, bio-reactor feeding, pilot plant facilities, pharmaceutical production and any application using flexible bio-bag
  • Disposable single cycle sleeve.
  • Save time and money on cleaning.
  • Eliminates dead legs where bacteria can grow.
  • Connects to most bio-bag connections.
  • Steam-through connections will transform your steam-in place (SIP) process by making the connections cleaner, faster and easier.
  • Innovative product design allows for a one-step process for sterilization and media transfer while eliminating dead spots, reducing the number of connections and eliminating the need for a laminar flow hood.
  • The terminations on the fittings are set to industry standards for bio-reactors and popular sizes of flexible tubing, speeding the adaptation to existing systems and processes.
  • Tear-away sleeve protects the fitting from actuation prior to complete sterilization.
  • The polysulfone body and silicone membrane materials meet requirements of USP Class VI, and physicochemical tests.
Specifications
Description Sanitary x Barb
Material Polysulfone
Tubing Size 3/4 in. x 1-1/2 in.
Hose Barb 3/4 in.
Outlet Port 1-1/2 in.
Sterilization Method Autoclave, gamma, or steam-in-place (SIP)
Product Type Sanitary Connector, PC Sleeve, Single Cycle x Barb Connector, 1/2 in.