Learn More
Cole-Parmer™ CPC™ Valved In-Line Coupling with Flared Fittings
Click to view available options
Diameter (English) Outer:
1/4 in.
Unit Size:
Each
Description
- ChemQuik quick-disconnect couplings require a fitting body and an insert of the same size (order separately).
- 100% metal-free construction.
- Single-handed operation.
- Valves for flow shut-off when disconnected.
- Make and break tubing connections without separating the tubing from the fitting.
- Create your own combinations for enhanced flexibility.
- Easily and intuitively switch lines to decrease downtime and increase efficiency.
- Reduce spills with valved fittings for additional safety.
- Select from various materials including plastic quick disconnects.
- Available with barbed, pipe, compression and more.
Specifications
Specifications
| Description | Quick-Disconnect x Flare |
| Material | Polypropylene |
| Diameter (English) Outer | 1/4 in. |
| Product Type | In-Line Coupling Body, flared fittings, valved, 1/4 in. OD, PP. |