missing translation for 'onlineSavingsMsg'
Learn More

CPD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-15979

Product Code. 18106721

  • € 564.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CPD Polyclonal specifically detects CPD in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CPD
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
carboxypeptidase D, GP180, metallocarboxypeptidase D
Rabbit
153 kDa
0.1 mL
Primary
Expected to cross react based on sequence identity: Rat (89%).
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot
1mg/mL
Western Blot 1:500-1:3000
O75976
CPD
Recombinant protein encompassing a sequence within the center region of human Carboxypeptidase D. The exact sequence is proprietary.
Affinity Purified
RUO
1362
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only