missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GST Epitope Tag Antibody, Alexa Fluor™ 405, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-325AF405
This item is not returnable.
View return policy
Description
GST Epitope Tag Polyclonal antibody specifically detects GST Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications
| GST Epitope Tag | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| Epitope Tag |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GSTM1 | |
| Goats were immunized with Glutathione-S-Transferase (GST) from Schistosoma japonicum. Antibody was isolated by affinity chromatography using GST immobilized on solid support. | |
| 0.1 mL | |
| Asthma, Cancer, Epitope Tags, GST Epitope Tags, Immunology | |
| 2944 | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction