missing translation for 'onlineSavingsMsg'
Learn More

CPI17 alpha Antibody [Alexa Fluor« 488], Novus Biologicals Biologicals™

Product Code. 30492917 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30492917 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30492917 Supplier Novus Biologicals Supplier No. NBP335103AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CPI17 alpha Polyclonal antibody specifically detects CPI17 alpha in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPI17 alpha
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias 17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human CPI17 alpha (NP_150281.1).,, Sequence:, MAAQRLGKRVLSKLQSPSRARGPGGSPGGLQKRHARVTVKYDRRELQRRLDVEKWIDGRLEELYRGMEADMPDEINIDELLELESEEERSRKIQGLLKSCGKPVEDFIQELLAKLQGLHRQPGLRQPSPSHDGSLSPLQDRARTAHP
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 94274
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.