Learn More
CPI17 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals H00094274-D01P
Product Code. 18474279
  • € 605.00 / 0.10mg
Description
CPI17 alpha Polyclonal antibody specifically detects CPI17 alpha in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CPI17 alpha
Polyclonal
PBS (pH 7.4)
17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A
PPP1R14A (NP_150281.1, 1 a.a. - 147 a.a.) full-length human protein. MAAQRLGKRVLSKLQSPSRARGPGGSPGGLQKRHARVTVKYDRRELQRRLDVEKWIDGRLEELYRGMEADMPDEINIDELLELESEEERSRKIQGLLKSCGKPVEDFIQELLAKLQGLHRQPGLRQPSPSHDGSLSPLQDRARTAHP
0.1 mg
Protein Phosphatase
94274
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
NP_150281.1
Rabbit
Protein A purified