missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CPLX1 Monoclonal Antibody (K40066_9A4), Invitrogen™

Product Code. p-200088640
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30210844 50 μL 50µg
30197367 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30210844 Supplier Invitrogen™ Supplier No. MA555624

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: GATKDMGKML GGDEEKDPDA AKKEEERQEA LRQAEEERKA KYAKMEAERE AVRQGIRDKY GIKKKEEREA EAQAAMEANS EGSLTRPKKA IPPGCGDEVE EEDESILDTV

CPLX1 encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPLX1
Applications Western Blot
Classification Monoclonal
Clone K40066_9A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Cplx1
Gene Accession No. O14810, P63040, P63041
Gene Alias 921-S; complexin 1; complexin I; Complexin-1; Cplx1; CPX I; CPX1; CPX-I; synaphin 2; synaphin-2
Gene Symbols Cplx1
Host Species Mouse
Immunogen Recombinant human CPLX1 (immunogen range is 11-120).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10815, 12889, 64832
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.