missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPLX4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05573-100ul
This item is not returnable.
View return policy
Description
CPLX4 Polyclonal antibody specifically detects CPLX4 in Rat samples. It is validated for Western Blot
Specifications
| CPLX4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CPLX4 complexin 4, CPXIV, CPX-IV | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human CPLX4 (NP_857637.1). MAFLMKSMISNQVKNLGFGGGSEENKEEGGASDPAAAQGMTREEYEEYQKQMIEEKMERDAAFTQKKAERACLRVHLREKYRLPKSEMDENQIQMAGDDVDLPEDLRKMVDEDQEEEEDKDSILGQIQNLQNMDLDTIKEKAQATFTEIKQTAEQKCSVM | |
| 100 μg | |
| Neuroscience | |
| 339302 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction