Learn More
CPM Mouse anti-Human, Clone: OTI2D11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807288
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.Specifications
| CPM | |
| Monoclonal | |
| Unconjugated | |
| CPM | |
| carboxypeptidase M | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1368 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P14384 | |
| CPM | |
| Human recombit protein fragment corresponding to amino acids 124-423 of human CPM produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.