missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPM Mouse anti-Human, Clone: OTI2D11, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807288S
This item is not returnable.
View return policy
Description
CPM Monoclonal antibody specifically detects CPM in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CPM | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P14384 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1368 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D11 | |
| Unconjugated | |
| CPM | |
| 1110060I01Rik; 5730456K23Rik; AA589379; Carboxypeptidase M; Cpm; E030045M14Rik; renal carboxypeptidase; urinary carboxypeptidase B | |
| Human recombinant protein fragment corresponding to amino acids 124-423 of human CPM produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction