missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPNE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144931
This item is not returnable.
View return policy
Description
Immunogen sequence: KNNLNPTWKR FSVPVQHFCG GNPSTPIQVQ CSDYDSDGS Antigen sequence identity: Mouse 87%; Rat 85%.
Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a calcium-dependent protein that also contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, the encoded protein does not contain a predicted signal sequence or transmembrane domains. This protein has a broad tissue distribution and it may function in membrane trafficking. This gene and the gene for RNA binding motif protein 12 overlap at map location 20q11.21. Alternate splicing results in multiple transcript variants encoding different proteins.
Specifications
| CPNE1 | |
| Polyclonal | |
| Unconjugated | |
| CPNE1 | |
| 1810028N16Rik; Chromobindin 17; copine 1; copine I; copine III; copine III, like; copine-1; COPN1; CPN1; Cpne1; cpne3; cpne3l; MGC1142; mKIAA4108; OTTHUMP00000030775; OTTHUMP00000030776; Unknown (protein for IMAGE:8126132); wu:fb69d07; wu:fc45a04; zgc:55873 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8904 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99829 | |
| CPNE1 | |
| Recombinant protein fragment of Human CPNE1 (Product #RP-104834) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction