missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPPED1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
€ 185.00 - € 405.00
Specifications
| Antigen | CPPED1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17960533
|
Invitrogen™
PA5145456 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17970263
![]() |
Invitrogen™
PA5145455 |
200 μL |
€ 405.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein phosphatase that dephosphorylates AKT family kinase specifically at 'Ser-473', blocking cell cycle progression and promoting cell apoptosis. May play an inhibitory role in glucose uptake by adipocytes.Specifications
| CPPED1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q66H71, Q8BFS6, Q9BRF8 | |
| 223978, 302890, 55313 | |
| Recombinant Mouse CPPED1 protein, Met1-Glu312 (Accession #Q8BFS6). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| C530044N13Rik; calcineurin like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain-containing protein 1; complete S transactivated protein 1; complete S-transactivated protein 1; Cpped1; CSTP1; RGD1306568; Serine/threonine-protein phosphatase CPPED1; Vcalcineurin-like phosphoesterase domain containing 1 | |
| CPPED1 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| CPPED1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title