missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPS1 Antibody (CPS1/8416), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21108AF350
This item is not returnable.
View return policy
Description
CPS1 Monoclonal antibody specifically detects CPS1 in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CPS1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16 | |
| Recombinant human CPS1 protein | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 1373 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| CPS1/8416 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction