missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
€ 164.00 - € 413.00
Specifications
Antigen | CPSF1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17773464
|
Invitrogen
PA5122069 |
20 μL |
€ 164.00
20µL |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17763464
|
Invitrogen
PA5122068 |
100 μL |
€ 413.00
100µL |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Midline-1 (Tripartite motif-containing protein 18, Putative transcription factor XPRF, RING finger protein 59) is a 667 amino acid protein encoded by the human gene MID1. Midline-1 belongs to the TRIM/RBCC family and contains two B box-type zinc fingers, one B30.2/SPRY domain, one COS domain, one fibronectin type-III domain and one RING-type zinc finger. Midline-1 is believed to have E3 ubiquitin ligase activity which targets the catalytic subunit of protein phosphatase 2 for degradation. It is a cytoplasmic protein found as a homodimer or heterodimer with Midline-2. It also interacts with IGBP1 (Lymphocyte signaling protein A4). Defects in MID1 are the cause of Opitz syndrome type I (OS-I). OS-I is an X-linked recessive disorder characterized by hypertelorism, genital-urinary defects such as hypospadias in males and splayed labia in females, lip-palate-laryngotracheal clefts, imperforate anus, developmental delay and congenital heart defects.Specifications
CPSF1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q10570 | |
29894 | |
Met1-Leu300 (Accession Q10570), with N-terminal His Tag | |
Primary | |
-20°C | |
Cpsf1 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
cleavage and polyadenylation specific factor 1; cleavage and polyadenylation specific factor 1, 160kDa; cleavage and polyadenylation specificity factor 1; cleavage and polyadenylation specificity factor 160 kDa subunit; Cleavage and polyadenylation specificity factor subunit 1; CPSF 160 kDa subunit; CPSF1; CPSF160; HSU37012; P/cl.18; polyadenylation specificity factor | |
Cpsf1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |