missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 sigma/Stratifin Antibody (CPTC-SFN-2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08214F
This item is not returnable.
View return policy
Description
14-3-3 sigma/Stratifin Monoclonal antibody specifically detects 14-3-3 sigma/Stratifin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications
| 14-3-3 sigma/Stratifin | |
| Monoclonal | |
| FITC | |
| 14-3-3 protein sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, stratifin, YWHAS | |
| Recombinant human full-length 14-3-3 sigma/Stratifin protein (Uniprot: P31947) | |
| 100 μg | |
| Cell Cycle and Replication | |
| 2810 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| CPTC-SFN-2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction