missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPSF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557005
This item is not returnable.
View return policy
Description
Immunogen sequence: PFNLLCYQLQ KLTGDVEELE IQEKPALKVF KNITVIQEPG MVVLEWLANP SNDMYADTVT TVILEVQSNP KIRKGAVQK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Most eukaryotic mRNA precursors (pre-mRNAs) undergo extensive maturational processing, including cleavage and polyadenylation at the 3-prime end. CPSF3 encodes the 73-kD subunit of the cleavage and polyadenylation specificity factor (CPSF) and is the pre-mRNA 3-prime-end-processing endonuclease (Mandel et al., 2006).
Specifications
| CPSF3 | |
| Polyclonal | |
| Unconjugated | |
| Cpsf3 | |
| cleavage and polyadenylation specific factor 3; cleavage and polyadenylation specific factor 3, 73kDa; cleavage and polyadenylation specificity factor 3; cleavage and polyadenylation specificity factor 73 kDa subunit; Cleavage and polyadenylation specificity factor subunit 3; CPSF; CPSF 73 kDa subunit; Cpsf3; Cpsf73; CPSF-73; mRNA 3'-end-processing endonuclease CPSF-73; YSH1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 298916, 51692, 54451 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9QXK7, Q9UKF6 | |
| Cpsf3 | |
| Recombinant protein corresponding to Human CPSF3. Recombinant protein control fragment (Product #RP-96050). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction