missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1B Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 213.00 - € 507.00
Specifications
| Antigen | CPT1B |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230810
|
Novus Biologicals
NBP3-33504-100ul |
100 μL |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229290
|
Novus Biologicals
NBP3-33504-20ul |
20 μL |
€ 213.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPT1B Monoclonal antibody specifically detects CPT1B in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| CPT1B | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 1375 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| carnitine O-palmitoyltransferase 1, muscle isoform, carnitine O-palmitoyltransferase I, mitochondrial muscle isoform, Carnitine O-palmitoyltransferase I, muscle isoform, Carnitine palmitoyltransferase 1B, carnitine palmitoyltransferase 1B (muscle), Carnitine palmitoyltransferase I-like protein, CPT I, CPT1M, CPT1-MMCCPT1, CPTI, CPTI-M, EC 2.3.1, EC 2.3.1.21, FLJ55729, FLJ58750, KIAA1670, MCPT1, M-CPT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 313-500 of human CPT1B (NP_004368.1).,, Sequence:, FNTTRIPGKDTDVLQHLSDSRHVAVYHKGRFFKLWLYEGARLLKPQDLEMQFQRILDDPSPPQPGEEKLAALTAGGRVEWAQARQAFFSSGKNKAALEAIERAAFFVALDEESYSYDPEDEASLSLYGKALLHGNCYNRWFDKSFTLISFKNGQLGLNAEHAWADAPIIGHLWEFVLGTDSFHLGYTE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title