missing translation for 'onlineSavingsMsg'
Learn More

CPT1B Monoclonal Antibody (OTI2A6), TrueMAB™, OriGene

Product Code. 17573874
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17573874 100 μL 100µL
17593874 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17573874 Supplier Origene Technologies Supplier No. TA810597

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and one of the transcripts is bicistronic.

Specifications

Antigen CPT1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A6
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CPT1B
Gene Accession No. Q92523
Gene Alias CPT1-M, CPT1M, CPTI, CPTI-M, M-CPT1, MCCPT1, MCPT1
Gene Symbols CPT1B
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 187-440 of human CPT1B produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1375
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.