missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1B Monoclonal Antibody (OTI6B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810598
This item is not returnable.
View return policy
Description
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and one of the transcripts is bicistronic.Specifications
| CPT1B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92523 | |
| CPT1B | |
| Human recombit protein fragment corresponding to amino acids 187-440 of human CPT1B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6B8 | |
| Unconjugated | |
| CPT1B | |
| CPT1-M, CPT1M, CPTI, CPTI-M, M-CPT1, MCCPT1, MCPT1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1375 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction