missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110953
This item is not returnable.
View return policy
Description
Immunogen sequence: MMVECSKYHG QLTKEAAMGQ GFDRHLFALR HLAAAKGIIL PELYLDPAYG QINHNVLSTS TLSSPAVNLG GFAPVVSDGF GVGYAVHDNW IGCNVSSYP Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 83%.
Carnitine palmitoyltransferase II precursor (CPT2) is a nuclear protein which is transported to the mitochondrial inner membrane. CPT2 together with carnitine palmitoyltransferase I oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders.
Specifications
| CPT2 | |
| Polyclonal | |
| Unconjugated | |
| CPT2 | |
| AI323697; Carnitine O-palmitoyltransferase 2, mitochondrial; carnitine palmitoyltransferase 2; carnitine palmitoyltransferase II; CPT II; CPT1; CPT2; Cpt-2; CPTASE; CPTII; IIAE4; OTTHUMP00000010493; testicular secretory protein Li 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1376 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P23786 | |
| CPT2 | |
| Recombinant Protein corresonding to Human CPT2. Recombinant protein control fragment (Product #RP-94317). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction