missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CPVL Polyclonal Antibody
GREENER_CHOICE

Product Code. 15761082 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15761082 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15761082 Supplier Invitrogen™ Supplier No. PA563308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LYQIGLLDEK QKKYFQKQCH ECIEHIRKQN WFEAFEILDK LLDGDLTSDP SYFQNVTGCS NYYNFLRCTE PEDQLYYVK Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 66%.

The protein encoded by this gene is a carboxypeptidase and bears strong sequence similarity to serine carboxypeptidases. Carboxypeptidases are a large class of proteases that act to cleave a single amino acid from the carboxy termini of proteins or peptides. The exact function of this protein, however, has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPVL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cpvl
Gene Accession No. Q9H3G5
Gene Alias 4933436L16Rik; carboxypeptidase WUG; carboxypeptidase, vitellogenic like; carboxypeptidase, vitellogenic-like; CP-Mac carboxypeptidase; CPVL; HVLP; probable serine carboxypeptidase CPVL; PSEC0124; tcag7.235; UNQ197/PRO223; VCP-like protein; vitellogenic carboxypeptidase-like protein; VLP
Gene Symbols Cpvl
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPVL. Recombinant protein control fragment (Product #RP-103258).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54504
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.