missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPZ Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 458.85 - € 700.35
Specifications
| Antigen | CPZ |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351376
|
Novus Biologicals
NBP3-17157-25UL |
25 μg |
€ 486.00 € 458.85 / 25µL Save € 27.15 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18368842
|
Bio-Techne
NBP3-17157-100UL |
100 μg |
€ 741.00 € 700.35 / 100µL Save € 40.65 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPZ Polyclonal antibody specifically detects CPZ in Human samples. It is validated for ImmunofluorescenceSpecifications
| CPZ | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| carboxypeptidase Z, EC 3.4.17, EC 3.4.17.-, EC 3.4.17.10, EC 3.4.17.3, MGC99682 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SYPFDFSKHPQEEKMFSPTPDEKMFKLLSRAYADVHPMMMDRSENRCGGNFLKRGSIINGADWYSFTGGMSDFNY | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 8532 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title