missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CR1 Monoclonal Antibody (OTI2A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811726S
This item is not returnable.
View return policy
Description
CR1 Monoclonal antibody specifically detects CR1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P00742 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2159 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A8 | |
| Unconjugated | |
| F10 | |
| C3 binding protein; C3b/C4b receptor; C3-binding protein; C3BR; C4BR; CD35; CD35 antigen; complement component (3b/4b) receptor 1 (Knops blood group); Complement Component (3b/4b) receptor 1 including Knops blood group system; complement component 3b/4b receptor 1 (Knops blood group); complement receptor type 1; CR1; KN; Knops blood group antigen | |
| Human recombinant protein fragment corresponding to amino acids 41-488 of human F10 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction