missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP1 Antibody (SR1172), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22148-50ul
This item is not returnable.
View return policy
Description
CRABP1 Monoclonal antibody specifically detects CRABP1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CRABP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1172 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| cellular retinoic acid binding protein 1, CRABP, CRABPI, CRABP-Icellular retinoic acid-binding protein 1, RBP5Cellular retinoic acid-binding protein I | |
| A synthesized peptide derived from human CRABP1 (Uniprot #: P29762) | |
| 50 μg | |
| Stem Cell Markers | |
| 1381 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction