missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRABP1 Monoclonal Antibody (C-1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA3813

Product Code. 11595782

  • € 554.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA3-813 detects cellular retinoic acid binding protein I (CRABPI) from human, mouse, monkey, feline, bovine, rabbit, rat, turtle and chicken tissues. This antibody does not react with chameleon, goldfish and frog. This antibody does not cross-react with cellular retinol binding protein (CRBP), performic acid oxidized CRBP, interphotoreceptor retinoid binding protein or retinol binding protein. MA3-813 has been successfully used in Western blot, immunohistochemistry and ELISA procedures. By Western blot, this antibody detects a 16 kDa protein representing CRABPI from rat retinal supernatant. Immunohistochemical staining of CRABPI in rat retina with MA3-813 results in its localization to amacrine somata and laminae in the inner plexiform layer. The MA3-813 antigen is oxidized CRABPI from bovine retina.

This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRABP1
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P29762, P40220, P62964, P62965, P62966
CRABP1
Oxidized CRABPI from bovine retina.
100 μL
Primary
Human, Mouse, Rat, Rabbit, Monkey, Bovine, Feline, Chicken, Reptile
Antibody
IgG2b
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
C-1
Unconjugated
CRABP1
AI326249; cCRABP-I; cellular retinoic acid binding protein 1; cellular retinoic acid binding protein I; cellular retinoic acid binding protein I (CRABP-I) homologue; cellular retinoic acid-binding protein 1; Cellular retinoic acid-binding protein I; cellular retinoic-acid-binding protein; CRABP; Crabp1; Crabp-1; CrabpI; CRABP-I; RBP5; Rbp-5; retinoic acid-binding protein
Mouse
Affinity chromatography, Protein A
RUO
100338607, 101090941, 12903, 1381, 25061, 282201, 374211
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.