missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP2 Antibody (OTI10D6), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70448PEATT594
This item is not returnable.
View return policy
Description
CRABP2 Monoclonal specifically detects CRABP2 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| CRABP2 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| CRABP2 | |
| Full length recombinant protein of human CRABP2 (NP_001869) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Flow Cytometry | |
| OTI10D6 | |
| Flow Cytometry | |
| cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 1382.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction