missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRABP2 Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
€ 185.00 - € 415.00
Specifications
| Antigen | CRABP2 |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17940763
![]() |
Invitrogen™
MA551145 |
200 μL |
€ 415.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17928732
|
Invitrogen™
MA551146 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.Specifica
| CRABP2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| AI893628; cellular lipid-binding protein; cellular retinoic acid binding protein 2; cellular retinoic acid binding protein II; cellular retinoic acid-binding protein 2; Cellular retinoic acid-binding protein II; Crabp2; Crabp-2; CrabpII; CRABP-II; RBP6; RP11-66D17.4 | |
| CRABP2 | |
| IgG2b κ | |
| Protein A/G | |
| CRABP2 |
| C1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig, Rat | |
| P29373, P51673 | |
| 100155151, 1382, 29563 | |
| Recombinant Human CRABP2 protein, Met1-Glu138 (Accession #P29373). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto