missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRACC/SLAMF7 Antibody (002), DyLight 680 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89961FR
This item is not returnable.
View return policy
Description
CRACC/SLAMF7 Monoclonal antibody specifically detects CRACC/SLAMF7 in Human samples. It is validated for ELISA
Specifications
| CRACC/SLAMF7 | |
| Monoclonal | |
| DyLight 680 | |
| 19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CRACC/SLAMF7 (NP_067004.3; Met1-Met226). | |
| 0.1 mL | |
| Immunology | |
| 57823 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 002 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction