Learn More
CRALBP Antibody (B2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-74392
This item is not returnable.
View return policy
Description
CRALBP Monoclonal specifically detects CRALBP in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.