missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Craniofacial Development Protein 1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18436AF350
This item is not returnable.
View return policy
Description
Craniofacial Development Protein 1 Polyclonal antibody specifically detects Craniofacial Development Protein 1 in Human samples. It is validated for Western Blot
Specifications
| Craniofacial Development Protein 1 | |
| Polyclonal | |
| Western Blot | |
| BCNTBUCENTAUR, Bucentaur, CP27phosphoprotein (Bucentaur), craniofacial development protein 1, p97, SWC5, Yeti | |
| Recognizes region between residues Between Between 249 and 299.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10428 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction