missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRB1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41201C
This item is not returnable.
View return policy
Description
CRB1 Polyclonal antibody specifically detects CRB1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| CRB1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DyLight 650 | |
| crumbs homolog 1 (Drosophila), LCA8 | |
| Antibody was raised against an 18 amino acid peptide near the center of human CRB1. The immunogen is located within amino acids 780 - 830 of CRB1. Amino Acid Squence: ISLKIKPYKIELYQSSQ | |
| 0.1 mL | |
| Signal Transduction | |
| 23418 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction