missing translation for 'onlineSavingsMsg'
Learn More

CRB1 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen CRB1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17581346
View Documents
Proteintech
26598-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17571346
View Documents
Proteintech
26598-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CRB1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CRB1. This antibody recognizes mouse antigen. The CRB1 antibody has been validated for the following applications: IHC, ELISA analysis.

Specifications

Specifications

CRB1
Polyclonal
Rabbit
23418
Primary
Immunohistochemistry
Unconjugated
Mouse
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.