missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRBN Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92446-0.1ml
This item is not returnable.
View return policy
Description
CRBN Polyclonal antibody specifically detects CRBN in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CRBN | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| cereblon, DKFZp781K0715, MGC27358, MRT2A, non-syndromic, autosomal recessive, 2A, protein cereblon, protein x 0001 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CRBN (NP_057386.2). MAGEGDQQDAAHNMGNHLPLLPAESEEEDEMEVEDQDSKEAKKPNIINFDTSLPTSHTYLGADMEEFHGRTLHDDDSCQVIPVLPQVMMILIPGQTLPLQLFHPQEVSMVRNLIQKDRTFAVLAYSNVQEREAQFGTTAEIYAYREEQDFGIEIVKVKAIGRQRFKVLELRTQSDGIQQAKVQILPECVLPSTMSAVQLESLNKCQIFPSKPVSREDQCSYKWWQKYQKRKFHCANLTSWPRWLYSLYDAETLMDRIKKQLREWDENLKDDSLPSNPIDF | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience | |
| 51185 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction