missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cre Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56133AF700
This item is not returnable.
View return policy
Description
Cre Polyclonal specifically detects Cre in Mouse, Cre/lox System samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Cre | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Cyclization recombinase | |
| A synthetic peptide made to a portion of bacteriophage protein CRE (within amino acids 50-100). [UniProt# P06956] | |
| 0.05 mL | |
| Primary | |
| Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2777477 | |
| Store at 4C in the dark. | |
| Cre/lox Systems |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction