missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase BB Antibody (2ba6), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59601JF549
This item is not returnable.
View return policy
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), Protein Array, CyTOF
Specifications
| Creatine Kinase BB | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Creatine Kinase Isoenzyme | |
| Human Creatine Kinase BB protein (Uniprot: P12277) | |
| 0.1 mL | |
| Cancer | |
| 1152 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 2ba6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction