missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase BB Antibody (CKBB/6565), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20318-100ug
This item is not returnable.
View return policy
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications
| Creatine Kinase BB | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Creatine Kinase Isoenzyme | |
| Recombinant human full-length Creatine Kinase BB protein | |
| 100 μg | |
| Cancer | |
| 1152 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| CKBB/6565 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction