missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Creatine Kinase BB Monoclonal Antibody (893CT29.1.1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA537618

Product Code. 17127208

  • € 510.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Creatine Kinase BB Monoclonal Antibody for Western Blot

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Creatine Kinase BB
Monoclonal
0.3 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P12277, Q04447
CKB
purified His-tagged CKB protein.
400 μL
Primary
Human, Mouse
Antibody
IgG1
Western Blot
893CT29.1.1
Unconjugated
CKB
Bck; B-CK; brain creatine kinase; Ck3; Ck-3; CKB; Ckbb; Ckbr; CPK-B; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29
Mouse
Protein G
RUO
1152, 12709
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.