missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Creatine Kinase BB Recombinant Rabbit Monoclonal Antibody (K01_3L24)
GREENER_CHOICE

Product Code. 30828586
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828586 100 μL
30825849 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828586 Supplier Invitrogen™ Supplier No. MA564385

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Creatine Kinase BB Recombinant Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Creatine Kinase BB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_3L24
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene CKB
Gene Accession No. P07335, P12277, Q04447
Gene Alias Bck; B-CK; brain creatine kinase; Ck3; Ck-3; CKB; Ckbb; Ckbr; CPK-B; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29
Gene Symbols CKB
Host Species Rabbit
Immunogen A synthetic peptide of human Creatine kinase B type.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1152, 12709, 24264
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.