missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase MB Antibody (1302 (1F2/1)), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62195CL1
This item is not returnable.
View return policy
Description
Creatine Kinase MB Monoclonal antibody specifically detects Creatine Kinase MB in Human samples. It is validated for ELISA,Radioimmune Assays
Specifications
| Creatine Kinase MB | |
| Monoclonal | |
| CoraFluor 1 | |
| Creatine Kinase Isoenzyme | |
| Highly purified human CKMB | |
| 0.1 mL | |
| metabolism | |
| 1152 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| ELISA, Radioimmune Assays (RIA) | |
| 1302 (1F2/1) | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction