missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine kinase MT 1A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05671-100ul
This item is not returnable.
View return policy
Description
Creatine kinase MT 1A Polyclonal antibody specifically detects Creatine kinase MT 1A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Creatine kinase MT 1A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Acidic-Type Mitochondrial Creatine Kinase, CKMT, CKMT1, CKMT1A, Creatine Kinase U-Type, Mitochondrial, Creatine Kinase, Mitochondrial 1 (Ubiquitous), Creatine Kinase, Mitochondrial 1A, EC 2.7.3.2, Mia-CK, Ubiquitous Mitochondrial Creatine Kinase, U-MtCK | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 258-417 of human Creatine kinase MT 1A (NP_001015001.1). FLIWVNEEDHTRVISMEKGGNMKRVFERFCRGLKEVERLIQERGWEFMWNERLGYILTCPSNLGTGLRAGVHIKLPLLSKDSRFPKILENLRLQKRGTGGVDTAATGGVFDISNLDRLGKSEVELVQLVIDGVNYLIDCERRLERGQDIRIPTPVIHTKH | |
| 100 μg | |
| Protein Kinase | |
| 548596 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction