missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase, Muscle/CKMM Antibody (CKMM/3341), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08304AF750
This item is not returnable.
View return policy
Description
Creatine Kinase, Muscle/CKMM Monoclonal antibody specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Creatine Kinase, Muscle/CKMM | |
| Monoclonal | |
| Alexa Fluor 750 | |
| CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK | |
| Recombinant full-length Creatine Kinase, Muscle/CKMM protein (Uniprot: P06732) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin), Protein Array | |
| CKMM/3341 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 1158 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction