missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CREB [p Ser133] Transcription Factor Assay Kit (Colorimetric)

Product Code. 18603310 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18603310 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18603310 Supplier Novus Biologicals™ Supplier No. KA1308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Assay Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Transcription Factor Assay Kit (Colorimetric)
Sample Type Nuclear Extracts
For Use With (Application) Functional
Gene ID (Entrez) 1385
Gene Symbol CREB1
Quantity 1 Kit
Research Discipline Phospho Specific
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.