missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB, p Ser271 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23250
This item is not returnable.
View return policy
Description
CREB Polyclonal antibody specifically detects CREB in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunofluorescence
Specifications
| CREB | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200 | |
| active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein | |
| Phospho-CREB (Ser-271) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Ser-271 in human CREB. This sequence is well conserved in rat, mouse, chicken, and zebrafish CREB. The site has some homology to the conserved site in ATF1 (Ser-198) and CREM (Ser-287). [UniProt# P16220] | |
| 0.1 mL | |
| Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
| 1385 | |
| Store at -20°C. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction