missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB3 regulatory factor Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06573MFV610
This item is not returnable.
View return policy
Description
CREB3 regulatory factor Polyclonal antibody specifically detects CREB3 regulatory factor in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| CREB3 regulatory factor | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| adult retina protein, C5orf41, chromosome 5 open reading frame 41, CREB3 regulatory factor, CREBRF, DKFZp313F2319, DKFZp686G2059, LRF, luman-recruiting factor | |
| Produced in rabbits immunized with E. coli-derived Human CREB3 regulatory factor fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 153222 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction