missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREBL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CREBL2 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858034
|
Proteintech
18007-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848034
|
Proteintech
18007-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
cAMP response element-binding protein-like-2 was identified in a search to find genes in a commonly deleted region on chromosome 12p13 flanked by ETV6 and CDKN1B genes, frequently associated with hematopoietic maligcies, as well as breast, non-small-cell lung and ovarian cancers. CREBL2 shares a 41% identity with CRE-binding protein over a 48-base long region which encodes the bZip domain of CREB. The bZip domain consists of about 30 amino acids rich in basic residues involved in DNA binding, followed by a leucine zipper motif involved in protein dimerization. This suggests that CREBL2 encodes a protein with DNA binding capabilities. The occurrence of CREBL2 deletion in maligcy suggests that CREBL2 may act as a tumor suppressor gene.Specifications
| CREBL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| O60519, Q32M00, Q5BJU6 | |
| 1389, 232430, 362453 | |
| CREBL2 Fusion Protein Ag12504 | |
| Primary | |
| -20°C | |
| CREBL2 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CREBL2 | |
| Crebl2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel